ContactUsLinkComponent
Monkey YKL-40/CHI3L1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of monkey YKL-40/CHI3L1 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid 1 * 96 Tests
Catalog # ANTIA326478-96
Supplier:  ANTIBODIES.COM
New Product
Monkey YKL-40/CHI3L1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of monkey YKL-40/CHI3L1 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid 1 * 96 Tests
Catalog # ANTIA326478-96
Supplier:  ANTIBODIES.COM