ContactUsLinkComponent
Mouse IL-1 beta Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-HRP, recombinant mouse IL-1&#0223 ELISA standard, and standard reconstitution buffer. This match 1 * 1.920 Tests
Catalog # ANTIA325339-2096
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-1 beta Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-HRP, recombinant mouse IL-1&#0223 ELISA standard, and standard reconstitution buffer. This match 1 * 1.920 Tests
Catalog # ANTIA325339-2096
Supplier:  ANTIBODIES.COM