ContactUsLinkComponent
SARS-CoV-2 SARS-CoV-2 Spike Glycoprotein S2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in v itro quantitative determination of sars-cov-2 SARS-CoV-2 Spike Glycoprotein S2 in serum, plasma, cell culture superna 1 * 96 Tests
Catalog # ANTIA326315-96
Supplier:  ANTIBODIES.COM
New Product
SARS-CoV-2 SARS-CoV-2 Spike Glycoprotein S2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in v itro quantitative determination of sars-cov-2 SARS-CoV-2 Spike Glycoprotein S2 in serum, plasma, cell culture superna 1 * 96 Tests
Catalog # ANTIA326315-96
Supplier:  ANTIBODIES.COM