ContactUsLinkComponent
Guinea Pig TGF beta 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of guinea pig TGF beta 1 in serum, platelet-poor plasma, tissue homogenates, cell culture supernates or oth 1 * 96 Tests
Catalog # ANTIA3231-96
Supplier:  ANTIBODIES.COM
New Product
Guinea Pig TGF beta 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of guinea pig TGF beta 1 in serum, platelet-poor plasma, tissue homogenates, cell culture supernates or oth 1 * 96 Tests
Catalog # ANTIA3231-96
Supplier:  ANTIBODIES.COM