ContactUsLinkComponent
Human NT-ProBNP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human NT-ProBNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 48 Tests
Catalog # ANTIA3217-48
Supplier:  ANTIBODIES.COM
New Product
Human NT-ProBNP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human NT-ProBNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 48 Tests
Catalog # ANTIA3217-48
Supplier:  ANTIBODIES.COM