ContactUsLinkComponent
Rat ProDynorphin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determ ination of rat ProDynorphin in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3775-96
Supplier:  ANTIBODIES.COM
New Product
Rat ProDynorphin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determ ination of rat ProDynorphin in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3775-96
Supplier:  ANTIBODIES.COM