Guinea Pig IL-10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of guinea pig IL-10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA3657-96
Supplier: ANTIBODIES.COM