ContactUsLinkComponent
Mouse APOA4/Apo-AIV ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of mouse APOA4/Apo-AIV in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other b 1 * 48 Tests
Catalog # ANTIA5538-48
Supplier:  ANTIBODIES.COM
New Product
Mouse APOA4/Apo-AIV ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of mouse APOA4/Apo-AIV in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other b 1 * 48 Tests
Catalog # ANTIA5538-48
Supplier:  ANTIBODIES.COM