Rat Slow Skeletal Myosin Heavy Chain ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of rat Slow Skeletal Myosin Heavy Chain in serum, plasma, tissue homogenates or other biolog 1 * 48 Tests
Artikelnr. ANTIA6498-48
Lieferant: ANTIBODIES.COM