ContactUsLinkComponent
Human IL-36 beta/IL-1F8 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human IL-36 beta/IL-1F8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or 1 * 48 Tests
Catalog # ANTIA6375-48
Supplier:  ANTIBODIES.COM
New Product
Human IL-36 beta/IL-1F8 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human IL-36 beta/IL-1F8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or 1 * 48 Tests
Catalog # ANTIA6375-48
Supplier:  ANTIBODIES.COM