ContactUsLinkComponent
Human IL-17RA Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human IL-17RA Receptor in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 96 Tests
Catalog # ANTIA7506-96
Supplier:  ANTIBODIES.COM
New Product
Human IL-17RA Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human IL-17RA Receptor in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 96 Tests
Catalog # ANTIA7506-96
Supplier:  ANTIBODIES.COM