Human cAMP Protein Kinase Catalytic Subunit alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for th e in vitro quantitative determination of human cAMP Protein Kinase Catalytic Subunit alpha in serum, plasma, tissue h 1 * 96 Tests
Catalog # ANTIA75751-96
Supplier: ANTIBODIES.COM