ContactUsLinkComponent
Rat Apolipoprotein CII/ApoC-II ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of rat Apolipoprotein CII/ApoC-II in serum, plasma, tissue homogenates, cell lysates, cell culture 1 * 96 Tests
Catalog # ANTIA6236-96
Supplier:  ANTIBODIES.COM
New Product
Rat Apolipoprotein CII/ApoC-II ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of rat Apolipoprotein CII/ApoC-II in serum, plasma, tissue homogenates, cell lysates, cell culture 1 * 96 Tests
Catalog # ANTIA6236-96
Supplier:  ANTIBODIES.COM