ContactUsLinkComponent
Human Carbonic Anhydrase 12/CA12 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Carbonic Anhydrase 12/CA12 in serum, plasma, tissue homogenates, cell lysates, cell cul 1 * 48 Tests
Catalog # ANTIA6991-48
Supplier:  ANTIBODIES.COM
New Product
Human Carbonic Anhydrase 12/CA12 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Carbonic Anhydrase 12/CA12 in serum, plasma, tissue homogenates, cell lysates, cell cul 1 * 48 Tests
Catalog # ANTIA6991-48
Supplier:  ANTIBODIES.COM