ContactUsLinkComponent
1 component: Chromium (III) (Cr 3+) 1000mg/l in Nitric Acid 2% CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # CMPLRM029049L1
New Product
1 component: Chromium (III) (Cr 3+) 1000mg/l in Nitric Acid 2% CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # CMPLRM029049L1