ContactUsLinkComponent
Powder Diffraction Line Profile Standard for Crystallite Size Analysis (Nano-Crystalline ZnO Powder) 1 * 3 g
Catalog # PROONIST-1979
New Product
Powder Diffraction Line Profile Standard for Crystallite Size Analysis (Nano-Crystalline ZnO Powder) 1 * 3 g
Catalog # PROONIST-1979