ContactUsLinkComponent
Revolution Microscope Upright & Inverted Capability Brightfield LED Illumination | LED Fluorescence Module | Multi-Dimensional Imaging: Time-Lapse | Multi-Point | Mosaic | Multi-Channel | Z-Stack | Motorized Epi- Fluorescence: 5 Channels | Mercury Fr 1 * 1 items
Catalog # ECHLRNSDEW81
Supplier:  DISCOVER ECHO
New Product
Revolution Microscope Upright & Inverted Capability Brightfield LED Illumination | LED Fluorescence Module | Multi-Dimensional Imaging: Time-Lapse | Multi-Point | Mosaic | Multi-Channel | Z-Stack | Motorized Epi- Fluorescence: 5 Channels | Mercury Fr 1 * 1 items
Catalog # ECHLRNSDEW81
Supplier:  DISCOVER ECHO