Micro titer plates, polystyrene, produced in low germ environment, with 96 wells, V-shape, max. 335 µl, 05-021-0100, 5 pcs./plastic bag, 100 pcs./carton 1 * 100 items
Catalog # MARI5200601
Supplier: MARIENFELD