Histamine Dihydrochloride [CAS:56-92-8] 12.5g/l in Water 10ml ampoule,, ,, CRM, ISO 17034 and ISO 17025,,,, Conservation 12 months.,,,, Store in a refrigerator at temperatures between 2°C to 8°C 1 * 2 Ampoul
Catalog # CMPLRD10385610.2
Supplier: CUSTOM MADE CHEMICALS LAB