NARROW NECKED BOTTLE LDPE TRANSP 50 ML 1 * 1 items
BURK0302-0050
: BURKLE
New Product
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Pipette stand, rotary
: BURKLE
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Transfer pipettes
: RATIOLAB
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection