ContactUsLinkComponent
vex athletic C XP * The uvex athletic C XP is a particularly lightweight cut protection glove for its cut protection class (level C). The micro-NBR foam coating is very flexible and breathable while also providing excellent grip and abrasion resistan 1 * 1 items
Catalog # UVEX60037
Supplier:  UVEX
New Product
vex athletic C XP * The uvex athletic C XP is a particularly lightweight cut protection glove for its cut protection class (level C). The micro-NBR foam coating is very flexible and breathable while also providing excellent grip and abrasion resistan 1 * 1 items
Catalog # UVEX60037
Supplier:  UVEX