ContactUsLinkComponent
Super Desiccator Wall, ACRYLIC/ALUMINIUM, 6 separate compartments , each compartment with: 1 elastic gasket-lined door with magnetic catch and cylinder lock, 1 flow meter for Regulation of gas flow, 1 desiccant tray, 1 mounted hygrometer, 1 removable 1 * 1 items
Catalog # BOHLV1996-08
Supplier:  Bohlender
New Product
undefined
Super Desiccator Wall, ACRYLIC/ALUMINIUM, 6 separate compartments , each compartment with: 1 elastic gasket-lined door with magnetic catch and cylinder lock, 1 flow meter for Regulation of gas flow, 1 desiccant tray, 1 mounted hygrometer, 1 removable 1 * 1 items
Catalog # BOHLV1996-08
Supplier:  Bohlender