ContactUsLinkComponent
ElectroReact Reduced Volume Complete Kit. Includes 6-position base plate, 6x reduced volume bottle reactors, 6x base plate adapters, 6x reactor caps, 12x banana plugs & 5x alligator clips. 1 * 1 KIT
Catalog # ASYNADS30-CK-RV
Supplier:  ASYNT
New Product
ElectroReact Reduced Volume Complete Kit. Includes 6-position base plate, 6x reduced volume bottle reactors, 6x base plate adapters, 6x reactor caps, 12x banana plugs & 5x alligator clips. 1 * 1 KIT
Catalog # ASYNADS30-CK-RV
Supplier:  ASYNT