FLOW CELL FOR PREP SFC 3 MM 1/16IN 1 * 1 items
Catalog # BUCH11076188
Supplier: BUCHI
New Product
Frequently Bought Together
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Formaldehyde solution Emprove Evolve 1 * 1 L
Supplier: MERCK PRODUCTION CHEMICALS
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Autoclave Bags, Polypropylene
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals