Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Precellys lysing kit - soil grinding sk38 (0.1mm glass beads,1.4mm zirconium oxide beads,1 glass bead of 4.0mm ) - 2 ml 1 * 50 KIT
Catalog # BERTP000915LYSK1A0
Supplier:
Bertin Instruments
New Product
Expand image
Precellys lysing kit - soil grinding sk38 (0.1mm glass beads,1.4mm zirconium oxide beads,1 glass bead of 4.0mm ) - 2 ml 1 * 50 KIT
Catalog # BERTP000915LYSK1A0
Supplier:
Bertin Instruments
New Product
Frequently Bought Together
PERFORM STERILE PAA -EU- 500 ML FL 1 * 10 Bottle
Supplier: SCHÜLKE & MAYR GMBH
Squalane 500ml pack 1 * 500 mL
Supplier: Apollo Scientific