ContactUsLinkComponent
Power contactor for eControl Lite (foreign vacuum cleaner) Vacuum cleaner with the following power specifications can be used: V 100 230 400 kW max 0,9 2,2 4(le=9A) Plug and socket must be provided and assembled by the customer. 1 * 1 items
Catalog # HAVE210037070
Supplier:  NEXOPART
New Product
Power contactor for eControl Lite (foreign vacuum cleaner) Vacuum cleaner with the following power specifications can be used: V 100 230 400 kW max 0,9 2,2 4(le=9A) Plug and socket must be provided and assembled by the customer. 1 * 1 items
Catalog # HAVE210037070
Supplier:  NEXOPART