Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
34 COMPONENTS: 100MG/L EACH IN HNO3 5% 1 * 100 mL
CMPLRM115455L1
:
CUSTOM MADE CHEMICALS LAB
New Product
34 COMPONENTS: 100MG/L EACH IN HNO3 5% 1 * 100 mL
CMPLRM115455L1
:
CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
VWR® Reagent Reservoirs
: VWR Collection
Pesticide standard, Imidacloprid, reference material, Honeywell Fluka™
: Honeywell Chemicals
Cole-Parmer® MSHM-200 Magnetic Stirring/Heating Mantles
: Cole-Parmer
Accessories for Binder KB, KBP PRO, KBF and KBF PRO
: Binder
VWR® Rigid qPCR Plates, 96-Well, with White Wells
: VWR Collection
Masterflex® Union Fittings, Hose Barb, Y-Connector, PVDF, Avantor®
: Avantor Fluid Handling