ContactUsLinkComponent
HygroGen with touch screen user interface, set-point control and programmer function, heated sample loop, 1x desiccant cell, 1x fill syringe, embedded HW4-P software and HC2-UAKS included. Order chamber door separately. 1 * 1 Each
Catalog # ROTRHG2-S
New Product
HygroGen with touch screen user interface, set-point control and programmer function, heated sample loop, 1x desiccant cell, 1x fill syringe, embedded HW4-P software and HC2-UAKS included. Order chamber door separately. 1 * 1 Each
Catalog # ROTRHG2-S