ContactUsLinkComponent
FREEZONE 6L -84C CONSOLE FREEZE DRYER, PTFE-COATED COLLECTOR, PURGE VALVE, STOPPERING TRAY DRYER, 230V, NATURAL REFRIGERATION GAS 1 * 1 items
Catalog # LBCP710626190
Supplier:  LABCONCO
New Product
FREEZONE 6L -84C CONSOLE FREEZE DRYER, PTFE-COATED COLLECTOR, PURGE VALVE, STOPPERING TRAY DRYER, 230V, NATURAL REFRIGERATION GAS 1 * 1 items
Catalog # LBCP710626190
Supplier:  LABCONCO