ContactUsLinkComponent
Orbital shaker-incubator with cooling with Bluetooth, 50 - 400 rpm, +4 to +80°C.  Order platform separately 1 * 1 items
Catalog # GRANES-20/80CEURO
Supplier:  GRANT INSTRUMENTS
New Product
Orbital shaker-incubator with cooling with Bluetooth, 50 - 400 rpm, +4 to +80°C.  Order platform separately 1 * 1 items
Catalog # GRANES-20/80CEURO
Supplier:  GRANT INSTRUMENTS