Reagecon Density Standard Premium Range 0.6878 g/mL at 25C 1 * 100 mL
REGEDEN25010PY
: REAGECON
New Product
Reagecon Density Standard Premium Range 0.6878 g/mL at 25C 1 * 100 mL
REGEDEN25010PY
: REAGECON
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection