ContactUsLinkComponent
Submersible pump set 90m, LDPE, 110V, US Submersible pump set, for purging and sampling monitoring wells to a depth of 90 m and inside diameter from 50 mm. Standard set with submersible pump Grundfos MP 1, converter, LDPE tube, cables, cooling jacket 1 * 1 ST
Catalog # EIJK1227SNUS
Submersible pump set 90m, LDPE, 110V, US Submersible pump set, for purging and sampling monitoring wells to a depth of 90 m and inside diameter from 50 mm. Standard set with submersible pump Grundfos MP 1, converter, LDPE tube, cables, cooling jacket 1 * 1 ST
Catalog # EIJK1227SNUS
Frequently Bought Together