Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Burette, automatic, 10:0.02ml, schellbach, without bottle, lateral stopcock, blue graduated, class AS, conformity certified 1 * 1 items
Catalog # HIRS354036008
Supplier:
HIRSCHMANN
New Product
Expand image
Burette, automatic, 10:0.02ml, schellbach, without bottle, lateral stopcock, blue graduated, class AS, conformity certified 1 * 1 items
Catalog # HIRS354036008
Supplier:
HIRSCHMANN
New Product
Frequently Bought Together
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Bulb pipettes (one mark), BLAUBRAND®, class AS
Supplier: Brand
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Supplier: VWR Collection
VWR® Burettes, with PTFE Key Stopcock
Supplier: VWR Collection
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection
VWR®, Bulb Pipettes, One Mark, Class AS
Supplier: VWR Collection
Bottles, Wide Neck with Screw Cap
Supplier: RIXIUS