2 COMPONENTS MIX 1 * 100 mL
Catalog # CMPLRM113852L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Rhodamine 6G ~95% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
cleaning fluid 5 Litre 1 * 5 L
Supplier: NEXOPART
TECHNOVIT 4004 LIQUIDE 500ML 1 * 1 ST
Supplier: HERAEUS KULZER
Acid red 27 ≥85-95% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Dehyd,Flex 100,4x1GAL 1 * 4 items
Supplier: Epredia
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
4-(2-Pyridylazo)resorcinol monosodim salt monohydrate, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY