HYDROQUINONE 0.1% IN METHANOL 1 * 10 Ampoul
Catalog # CMPLRD1129835.10
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
HYDROQUINONE 0.1% IN METHANOL 1 * 10 Ampoul
Catalog # CMPLRD1129835.10
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Sodium hypochlorite (14% Cl₂) in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator strips for coloured samples
Supplier: VWR Chemicals
Test kits, chlorine, Spectroquant®, Supelco®
Supplier: Merck
VWR®, Reagents in tablet form for photometric determination of chlorine
Supplier: VWR Chemicals
Test strips for nickel, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Standards, Spectroquant® CombiCheck, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Alcometer fuel cell breathalyser
Supplier: SURESCREEN