Orders LinkContactUsLinkComponent
Objectives
Plan semi apochromat phase objective, —
Catalog # OLYMN6410500
Supplier:   OLYMPUS OPTICAL
Objectives
Catalog # OLYMN6410500
Supplier:  OLYMPUS OPTICAL
Objectives
Objectives
Plan semi apochromat phase objective, —
Price
514,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    10X/0,3
  • Description:
    Plan semi apochromat phase objective
  • Seq:
    03
  • Working distance: