Orders LinkContactUsLinkComponent
Flasks, round bottom, with three parallel necks
Flasks, round bottom, with three parallel necks
100 ml, 14/23 NS, 14/23 NS
Catalog # NEUB0760-0100-14
Flasks, round bottom, with three parallel necks
Catalog # NEUB0760-0100-14
Flasks, round bottom, with three parallel necks
100 ml, 14/23 NS, 14/23 NS
Price
108,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Specifications
  • Capacity:
    100 ml
  • Seq:
    02
  • Side necks:
    14/23 NS
  • Centre neck:
    14/23 NS