Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer Capsules

Buffer Capsules

pH buffer capsules, GPH 4, amount: 5 capsules, 4,0, Red, 1 SET
Catalog # GRSG602614
Supplier:   Greisinger
Buffer Capsules
Catalog # GRSG602614
Supplier:  Greisinger
CAS Number:  
Buffer Capsules

pH buffer capsules, GPH 4, amount: 5 capsules, 4,0, Red, 1 SET

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 SET
Product Details & Documents

Cost-effective alternative: Buffer capsules and working sets. With buffer capsules, you can produce a 100 ml buffer solution in the most cost-effective manner. A particular advantage of the capsules is that their storage life is several years longer than that of ready-to-use solutions.

The capsules are offered in a practical working set: The most cost-effective entry with everything you need for a successful start in pH measurement, including cleaning solution and KCL100 storage/replacement electrolyte.

Specifications

Specifications

  • Pk:
    1 SET
  • Colour:
    Red
  • Description:
    pH buffer capsules, GPH 4, amount: 5 capsules
  • pH value:
    4,0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.