,,12 components: 100ug/ml each of B Ge Mo Nb P Re Si S Ta Ti W Zr in HNO3 5% HFtr ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM083308L1
Supplier: CUSTOM MADE CHEMICALS LAB