Label Rolls, Core2Write
Core2Write label, non-sterile, White, 50,8×25,4 mm, 1000 labels/roll, 4 rolls/case
VELTC2WR2X1BLK01
Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures. All labels can be customised or left blank.
- Always retain critical documentation, pre-printed and hand-written
- Extremely durable
- Abrasion and tear resistant
- Smear resistant
- Can withstand multiple chemicals
- Can withstand a wide temperature range (–70 to 200 ℃)
- Fully customisable by size, color, pre-printing
All roll labels come with a three inch plastic core. Can be applied to all types of surfaces: glass, poly, stainless steel. Can be autoclaved.
: Also available in sheets (sold separately).
- Colour:White
- Description:Core2Write label, non-sterile
- Dimensions:50,8×25,4 mm
- Seq:01
- Packed:1000 labels/roll, 4 rolls/case



