ContactUsLinkComponent
This alpha Amylase Activity Assay Kit is a quantitative assay kit for measuring alpha Amylase activity in serum, plasma, saliva, ani mal tissues, and plant tissues (seeds or newly germinated seedings). 1 * 96 Tests
Catalog # ANTIA319757-96
Supplier:  ANTIBODIES.COM
New Product
This alpha Amylase Activity Assay Kit is a quantitative assay kit for measuring alpha Amylase activity in serum, plasma, saliva, ani mal tissues, and plant tissues (seeds or newly germinated seedings). 1 * 96 Tests
Catalog # ANTIA319757-96
Supplier:  ANTIBODIES.COM