Orders LinkContactUsLinkComponent
Packaging and Storage Boxes, PP
Packaging and Storage Boxes, PP
Natural, 37×37×10 mm, PP
Catalog # SEMA7325
Supplier:   SEMADENI
Packaging and Storage Boxes, PP
Catalog # SEMA7325
Supplier:  SEMADENI
Packaging and Storage Boxes, PP
Natural, 37×37×10 mm, PP
Price
0,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
High-quality, elegant, robust cases with snap fasteners integrated into the contour and durable hinges.
  • Natural colour/translucent
  • Excellent printing and labelling options
The cases are perfect for product presentation thanks to the polished lid surfaces. Also ideal for packaging, transportation and storage of small parts, tools, writing materials, etc.
Specifications
  • Colour:
    Natural
  • Material:
    PP
  • Seq:
    01
  • W×D×H:
    37×37×10 mm