ContactUsLinkComponent
PRIME-XV Adipogenic Differentiation SFM is a serum-free complete medium optimized for the differentiation of human MSCs. This product does not contain antibiotics.Designed and manufactured to facilitate transfer from research to clinic: FDA, Federal, 1 * 100 mL
Catalog # SHBT91137
New Product
PRIME-XV Adipogenic Differentiation SFM is a serum-free complete medium optimized for the differentiation of human MSCs. This product does not contain antibiotics.Designed and manufactured to facilitate transfer from research to clinic: FDA, Federal, 1 * 100 mL
Catalog # SHBT91137