ContactUsLinkComponent
PRIME-XV T Cell CDM is the first commercially-available, chemically defined, animal component-free medium for the expansion and transduction of human T cells. The formulation is optimized to deliver consistently vigorous growth while maintaining T ce 1 * 1 L
Catalog # SHBT91154
New Product
PRIME-XV T Cell CDM is the first commercially-available, chemically defined, animal component-free medium for the expansion and transduction of human T cells. The formulation is optimized to deliver consistently vigorous growth while maintaining T ce 1 * 1 L
Catalog # SHBT91154