ContactUsLinkComponent
Chair Chrome, 5-branch, scuff-resistant, reinforced base with heavy-duty antistatic castors. Chrome gas-lift column with height adjustment from 470 to 605 mm and shock absorber. Deformable seat in antistatic black KANGO soft polyurethane, tilted of 2 1 * 1 items
Catalog # BIEN7EP91GBLN00905
Supplier:  Kango
New Product
Chair Chrome, 5-branch, scuff-resistant, reinforced base with heavy-duty antistatic castors. Chrome gas-lift column with height adjustment from 470 to 605 mm and shock absorber. Deformable seat in antistatic black KANGO soft polyurethane, tilted of 2 1 * 1 items
Catalog # BIEN7EP91GBLN00905
Supplier:  Kango