AMMONIUM OXALATE SOLUTION 1 * 100 mL
Catalog # CMPL1005901.L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Azure B (azure I) ≥85% (by HPLC), prepared by direct synthesis, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Potassium hydroxide ≥85% ACS (max. 2% K₂CO₃)
Supplier: Thermo Fisher Scientific
SCHIFF REAGENT 500ML 1 * 1 items
Supplier: Epredia
Fast Dark Blue R Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Methyl green zinc chloride salt <0,5% crystal violet 80% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond