POTASSIUM IODOPLATINATE TS USP TEST SOL. 1 * 50 mL
Catalog # SPCMP-236-50ML
Supplier: Spectrum Chemical
New Product
POTASSIUM IODOPLATINATE TS USP TEST SOL. 1 * 50 mL
Catalog # SPCMP-236-50ML
Supplier: Spectrum Chemical
New Product
Frequently Bought Together
Test strips, ascorbic acid, Reflectoquant®, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Sodium perchlorate monohydrate ≥99.0%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Face Masks, 3 ply, Non Woven PP
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® Erlenmeyer Flasks, with Standard Ground Joint
Supplier: VWR Collection
VWR® PESTINORM®, Acetonitrile, Secondary Reference Standard for GC
Supplier: VWR Chemicals