,,9 components: 1000mg/l each of Al Fe Mn Mg Ca Na K P Sc in HNO3 5% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at temperatures between 15C to 25C,,,, 1 * 100 mL
Catalog # CMPLRM109670L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together