,,10 components: 100mg/l each of As Cd Co Cr Cu Hg Ni Pb Se Zn in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months. (12 months after opening),, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM096012L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together