Orders LinkContactUsLinkComponent
Accessories for Continuous Level Control, SymLine®
Accessories for Continuous Level Control, SymLine®
Built-in signal box display, brushed stainless steel, 2 mm wall thickness, for signal boxes 1/2/5 with cable connection on the back (Version 2024)
Catalog # SCTV106733
Supplier:   S.C.A.T.
Accessories for Continuous Level Control, SymLine®
Catalog # SCTV106733
Supplier:  S.C.A.T.
Accessories for Continuous Level Control, SymLine®
Accessories for Continuous Level Control, SymLine®
Built-in signal box display, brushed stainless steel, 2 mm wall thickness, for signal boxes 1/2/5 with cable connection on the back (Version 2024)
Price
58,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
LED's and acoustic alarm warn you in time before the fill level of your collection containers reaches the critical level. With mute / snooze function and interface for controlling external devices. For installation in your media line of the fume cupboard.
  • Built-in Signalbox2 for Safety Waste Cap and other electronical level controlers
  • Touchless alarm-resetting fields for two containers
  • Two switch inputs and two switch outputs for peripheral units
  • Incl. 230 V power supply cable with open end and clamp
Suitable e.g. for equipment from Wesemann, Waldner, Bense, Hemling, Köttermann, Kewaunee, Carlo Erba, Modus Complete, Delagrave, and many more.
Specifications
  • Description:
    Built-in signal box display, brushed stainless steel, 2 mm wall thickness, for signal boxes 1/2/5 with cable connection on the back (Version 2024)
  • Seq:
    6